MilliporeSigma
  • β-1,3-Glucan recognition protein (βGRP) is essential for resistance against fungal pathogen and opportunistic pathogenic gut bacteria in Locusta migratoria manilensis.
β-1,3-Glucan recognition protein (βGRP) is essential for resistance against fungal pathogen and opportunistic pathogenic gut bacteria in Locusta migratoria manilensis.
Developmental and comparative immunology (2011-11-09)
Xiaoli Zheng, Yuxian Xia
ABSTRACT
Pattern recognition proteins, which form part of the innate immune system, initiate host defense reactions in response to pathogen surface molecules. The pattern recognition protein β-1,3-glucan recognition protein (βGRP) binds to β-1,3-glucan on fungal surfaces to mediate melanization via the prophenoloxidase (PPO)-activating cascade. In this study, cDNA encoding a 53-kDa βGRP (LmβGRP) was cloned from Locusta migratoria manilensis. LmβGRP mRNA shown to be constitutively expressed specifically in hemocytes and was highly upregulated following fungal infection. LmβGRP-silenced (LmβGRP-RNAi) mutant locusts exhibited significantly reduced survival rate following fungal infection (Metarhizium acridum) compared with the wild-type. Furthermore, LmβGRP-RNAi mutants exhibited abnormally loose stools indicative of a gut defect. 16S rRNA gene analysis detected the opportunistic pathogenic bacterium, Vibrio vulnificus in LmβGRP mutant but not wild-type locusts, suggesting changes in the composition of gut bacterial communities. These results indicate that LmβGRP is essential to gut immunity in L. migratoria manilensis.
Sigma-Aldrich
β-1,3-Glucan from Euglena gracilis