MilliporeSigma
  • Intra-coronal bleaching in young permanent and primary tooth with biologic perspectives.
Intra-coronal bleaching in young permanent and primary tooth with biologic perspectives.
The Journal of clinical pediatric dentistry (2011-11-04)
Divya S Sharma, Khushboo Barjatya, Anuradha Agrawal
ABSTRACT
The odd attracts society in odd manner, as is the case when a patient with discolored tooth smiles. Because of that, pediatric patients have psychological impact. Trauma and pulpal necrosis are the most common causes for discoloration of teeth. If tooth is intact, intra-coronal bleaching is the most conservative and noninvasive treatment modality provided, it is done cautiously. This article intends to present two case-reports of successful intra-coronal bleaching using milder (sodium perborate) and tissue-friendly bleaching agent with walking bleach.
Sigma-Aldrich
Sodium perborate tetrahydrate, 9-11% available Oxygen
Sigma-Aldrich
Sodium perborate tetrahydrate, purum p.a., 96% (RT)
Sigma-Aldrich
Sodium perborate monohydrate, powder, 20-100 mesh