MilliporeSigma
  • Colour stability of relined dentures after chemical disinfection. A randomised clinical trial.
Colour stability of relined dentures after chemical disinfection. A randomised clinical trial.
Journal of dentistry (2011-11-03)
Eduardo Buozi Moffa, Eunice Teresinha Giampaolo, Fernanda Emiko Izumida, Ana Cláudia Pavarina, Ana Lúcia Machado, Carlos Eduardo Vergani
ABSTRACT
This randomised clinical study evaluated the effect of chemical disinfection with sodium perborate or chlorhexidine on the colour stability of a hard chairside reline resin during six months. Hard chairside reline resin (Tokuyama Rebase Fast II) was used to reline complete dentures. After baseline colour measurements, the patients were randomly divided into 3 groups (n = 15) and allocation was concealed with the use of the BioStat program. The dentures were cleansed according to three methods: CG (control group)--brushing with coconut soap and soft toothbrush, PG (Perborate group)--brushing according to previous methods and disinfection with warmed sodium perborate solution (Corega Tabs) for 5 min, once a day for 6 months and ChxG (Chlorhexidine Group)--brushing according to CG and disinfection with chlorhexidine digluconate 2% for 5min once a day for 6 months. The data of ΔE*, ΔL*, Δa* and Δb* were analysed by 2-way repeated-measures ANOVAs and Tukey tests (α = 0.05). There were significant differences amongst groups for ΔL, Δa and Δb. The time had a significant effect on ΔE and ΔL, for all groups evaluated. Changes in some colour parameters (ΔL, Δa and Δb) of the reline resin Tokuyama Rebase were observed when the dentures were disinfected by perborate and chlorhexidine digluconate 2% solutions. The colour stability of was also influenced by time, regardless of disinfection or nondisinfection. Colour stability of the denture materials is one variable to be considered when choosing disinfection methods. The data in this study will be useful to clinicians when they are selecting disinfectant solutions for disinfection of relined denture.
Sigma-Aldrich
Sodium perborate monohydrate, powder, 20-100 mesh
Sigma-Aldrich
Sodium perborate tetrahydrate, purum p.a., 96% (RT)
Sigma-Aldrich
Sodium perborate tetrahydrate, 9-11% available Oxygen