MilliporeSigma
Properties of compatible solutes in aqueous solution.
Biophysical chemistry (2011-10-22)
Jens Smiatek, Rakesh Kumar Harishchandra, Oliver Rubner, Hans-Joachim Galla, Andreas Heuer
ABSTRACT
We have performed Molecular Dynamics simulations of ectoine, hydroxyectoine and urea in explicit solvent. Special attention has been spent on the local surrounding structure of water molecules. Our results indicate that ectoine and hydroxyectoine are able to accumulate more water molecules than urea by a pronounced ordering due to hydrogen bonds. We have validated that the charging of the molecules is of main importance resulting in a well defined hydration sphere. The influence of a varying salt concentration is also investigated. Finally we present experimental results of a DPPC monolayer phase transition that validate our numerical findings.
Sigma-Aldrich
Ectoine, ≥95.0% (HPLC)
Sigma-Aldrich
Ectoine, osmoprotectant