MilliporeSigma
  • Asymmetric reduction of ketones with recombinant E. coli whole cells in neat substrates.
Asymmetric reduction of ketones with recombinant E. coli whole cells in neat substrates.
Chemical communications (Cambridge, England) (2011-10-19)
Andre Jakoblinnert, Radoslav Mladenov, Albert Paul, Fabrizio Sibilla, Ulrich Schwaneberg, Marion B Ansorge-Schumacher, Pablo Domínguez de María
ABSTRACT
The asymmetric reduction of ketones is performed by using lyophilized whole cells in neat substrates with defined water activity (a(w)). Ketones and alcohols prone to be unstable in aqueous media can now be converted via biocatalysis.
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, ≥98%, FG