MilliporeSigma
  • Magnetic-fluorescent nanohybrids of carbon nanotubes coated with Eu, Gd co-doped LaF3 as a multimodal imaging probe.
Magnetic-fluorescent nanohybrids of carbon nanotubes coated with Eu, Gd co-doped LaF3 as a multimodal imaging probe.
Journal of colloid and interface science (2011-10-19)
Bingdi Chen, Hui Zhang, Ning Du, Bo Zhang, Yulian Wu, Donglu Shi, Deren Yang
ABSTRACT
Multifunctional magnetic-fluorescent nanohybrids were successfully fabricated by a facile layer-by-layer (LBL) self-assembly of in situ generated Eu, Gd co-doped LaF(3) nanocrystals (LaF(3):Eu:Gd) on the surface of multi-walled carbon nanotubes (MWNTs). Photoluminescence (PL) quenching occurred when LaF(3):Eu:Gd nanoparticles were directly coated on the surface of MWNTs. By growth of a SiO(2) shell spacer between MWNTs and LaF(3):Eu:Gd nanocrystals, we circumvented the PL quenching and achieved the magnetic-fluorescent MWNTs. Moreover, the nanohybrids showed powerful T(1) and T(2)-weighted magnetic resonance imaging (MRI) signal in water and could be used as MRI contrast agents. As a result, the nanohybrids can be expected to act as a promising multimodal MRI/optical imaging probe.
Sigma-Aldrich
Lanthanum(III) fluoride, anhydrous, powder, 99.99% trace metals basis