MilliporeSigma
Postmortem chemistry update part II.
International journal of legal medicine (2011-10-11)
Cristian Palmiere, Patrice Mangin
ABSTRACT
As a continuation of "Postmortem Chemistry Update Part I," Part II deals with molecules linked to liver and cardiac functions, alcohol intake and alcohol misuse, myocardial ischemia, inflammation, sepsis, anaphylaxis, and hormonal disturbances. A very important array of new material concerning these situations had appeared in the forensic literature over the last two decades. Some molecules, such as procalcitonin and C-reactive protein, are currently researched in cases of suspected sepsis and inflammation, whereas many other analytes are not integrated into routine casework. As in part I, a literature review concerning a large panel of molecules of forensic interest is presented, as well as the results of our own observations, where possible.
Sigma-Aldrich
Diethyl sulfate, 98%