MilliporeSigma
  • Light triggered detection of aminophenyl phosphate with a quantum dot based enzyme electrode.
Light triggered detection of aminophenyl phosphate with a quantum dot based enzyme electrode.
Journal of nanobiotechnology (2011-10-11)
Waqas Khalid, Gero Göbel, Dominik Hühn, Jose-Maria Montenegro, Pilar Rivera-Gil, Fred Lisdat, Wolfgang J Parak
ABSTRACT
An electrochemical sensor for p-aminophenyl phosphate (pAPP) is reported. It is based on the electrochemical conversion of 4-aminophenol (4AP) at a quantum dot (QD) modified electrode under illumination. Without illumination no electron transfer and thus no oxidation of 4AP can occur. pAPP as substrate is converted by the enzyme alkaline phosphatase (ALP) to generate 4AP as a product. The QDs are coupled via 1,4-benzenedithiol (BDT) linkage to the surface of a gold electrode and thus allow potential-controlled photocurrent generation. The photocurrent is modified by the enzyme reaction providing access to the substrate detection. In order to develop a photobioelectrochemical sensor the enzyme is immobilized on top of the photo-switchable layer of the QDs. Immobilization of ALP is required for the potential possibility of spatially resolved measurements. Geometries with immobilized ALP are compared versus having the ALP in solution. Data indicate that functional immobilization with layer-by-layer assembly is possible. Enzymatic activity of ALP and thus the photocurrent can be described by Michaelis- Menten kinetics. pAPP is detected as proof of principle investigation within the range of 25 μM-1 mM.
Sigma-Aldrich
Benzene-1,4-dithiol, 99% (GC)
Supelco
4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
4-Aminophenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Aminophenol, ≥98%
Sigma-Aldrich
Biphenyl-4,4′-dithiol, 95%