MilliporeSigma
  • Diastereocontrol in asymmetric allyl-allyl cross-coupling: stereocontrolled reaction of prochiral allylboronates with prochiral allyl chlorides.
Diastereocontrol in asymmetric allyl-allyl cross-coupling: stereocontrolled reaction of prochiral allylboronates with prochiral allyl chlorides.
Journal of the American Chemical Society (2011-10-07)
Laura A Brozek, Michael J Ardolino, James P Morken
ABSTRACT
Palladium-catalyzed allyl-allyl cross-coupling was investigated with substituted prochiral allylic boronates. These reactions deliver products bearing adjacent stereocenters, and the issue of diastereocontrol is therefore paramount. Under appropriately modified conditions, this allyl-allyl coupling strategy was found to apply to a range of substrates, generally occurring with high enantioselectivity (92:8 to >99:1 er) and good diastereoselection (4:1 to 14:1 dr).
Sigma-Aldrich
Allyl chloride, reagent grade, 98%
Sigma-Aldrich
Allyl chloride, ReagentPlus®, 99%