MilliporeSigma
  • Breakthrough in concentration quenching threshold of upconversion luminescence via spatial separation of the emitter doping area for bio-applications.
Breakthrough in concentration quenching threshold of upconversion luminescence via spatial separation of the emitter doping area for bio-applications.
Chemical communications (Cambridge, England) (2011-10-05)
Xiaomin Liu, Xianggui Kong, Youlin Zhang, Langping Tu, Yu Wang, Qinghui Zeng, Chunguang Li, Zhan Shi, Hong Zhang
ABSTRACT
The concentration quenching threshold of upconversion luminescence was broken through for the first time via a designed strategy: spatial separation of the emitter doping area.
Sigma-Aldrich
2,3-Benzofuran, 99%