MilliporeSigma
  • Synthesis of novel 6'-spirocyclopropyl-5'-norcarbocyclic adenosine phosphonic Acid analogues as potent anti-hiv agents.
Synthesis of novel 6'-spirocyclopropyl-5'-norcarbocyclic adenosine phosphonic Acid analogues as potent anti-hiv agents.
Nucleosides, nucleotides & nucleic acids (2011-10-05)
Lian Jin Liu, Eunae Kim, Joon Hee Hong
ABSTRACT
Novel 5'-norcarbocyclic adenosine phosphonic acid analogues with 6'-electropositive moiety such as spirocyclopropane were designed and synthesized from the commercially available diethylmalonate 5. Regioselective Mitsunobu reaction proceeded in the presence of an allylic functional group at a low reaction temperature in polar cosolvent [dimethylformamide (DMF)/1,4-dioxane] to give purine analogue 15. To improve cellular permeability and enhance the anti-human immunodeficiency virus (HIV) activity of this phosphonic acid, a SATE phosphonodiester nucleoside prodrug 23 was prepared. The synthesized adenosine phosphonic acids analogues 18-21 and 23 were subjected to antiviral screening against HIV-1.
Sigma-Aldrich
Diethyl malonate, ≥98%, FG
Sigma-Aldrich
Diethyl malonate, ReagentPlus®, 99%