MilliporeSigma
  • Fatigue resistance of acrylic resin denture base material reinforced with E-glass fibres.
Fatigue resistance of acrylic resin denture base material reinforced with E-glass fibres.
Gerodontology (2011-10-01)
Ozlem Gurbuz, Idil Dikbas, Fatma Unalan
ABSTRACT
The purpose of the study was to determine the effect of different forms and concentrations (2.5, 3, 4, 5% by volume) of glass fibres (chopped strand mat, continuous and woven) on fatigue resistance of acrylic denture base resin. The fatigue resistance was measured by applying repeated three-point bending deflection to the specimens, the cycle frequency of 1.05 g and magnitude of deflection of 2.0 mm. The number of loading cycles needed to cause a fracture in the test specimen was considered the fatigue resistance of the specimen. The results of this study revealed that the addition of three different glass fibre forms at all concentrations to acrylic resin did not produce a statistically significant increase in the fatigue resistance (p ≥ 0.05). This study also revealed that there were significant differences (p < 0.05) between glass fibres forms used concerning the effects on the fatigue resistance. This study showed that the woven glass fibres had a definite superiority over the chopped fibres and the continuous fibres in regard to the fatigue resistance of the acrylic denture base resin.
Sigma-Aldrich
Barium oxide, 97%
Sigma-Aldrich
Barium oxide, 99.99% trace metals basis