MilliporeSigma
  • Comparison of the course of biomarker changes and kidney injury in a rat model of drug-induced acute kidney injury.
Comparison of the course of biomarker changes and kidney injury in a rat model of drug-induced acute kidney injury.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2011-10-01)
Daisuke Sasaki, Atsushi Yamada, Hitomi Umeno, Hiroshi Kurihara, Shunji Nakatsuji, Shiro Fujihira, Kenjiro Tsubota, Mihoko Ono, Akira Moriguchi, Kouji Watanabe, Jiro Seki
ABSTRACT
To aid in evaluating the performance of biomarkers, we measured kidney injury biomarkers in rat models of drug-induced acute kidney injury. Rats were treated with site-specific nephrotoxins, puromycin, gentamicin, cisplatin, or 2-bromoethylamine. Fifteen biomarkers (β-2-microglobulin, calbindin, clusterin, cystatin-C, KIM-1, GST-α, GST-μ, NGAL, osteopontin, EGF, TIMP-1, VEGF, albumin, RPA-1, and urinary total protein) were examined in comparison with BUN, serum creatinine, and NAG. Some biomarkers, which were different depending in each nephrotoxin, showed ability to detect the prodromal stage of drug-induced kidney injury. Characteristic changing patterns of biomarkers were also found depending on the specific lesion site in the kidney. These data suggested that establishment of a suitable biomarker panel would facilitate detection of site-specific kidney injury with high sensitivity.
Sigma-Aldrich
2-Bromoethylamine hydrobromide, purum, ≥97.0% (AT)
Sigma-Aldrich
2-Bromoethylamine hydrobromide, 99%