MilliporeSigma
  • Differentiation of protonated aromatic regioisomers related to lignin by reactions with trimethylborate in a Fourier transform ion cyclotron resonance mass spectrometer.
Differentiation of protonated aromatic regioisomers related to lignin by reactions with trimethylborate in a Fourier transform ion cyclotron resonance mass spectrometer.
Journal of the American Society for Mass Spectrometry (2011-09-29)
Jayalakshmi Somuramasami, Penggao Duan, Lucas M Amundson, Enada Archibold, Brian E Winger, Hilkka I Kenttämaa
ABSTRACT
Several lignin model compounds were examined to test whether gas-phase ion-molecule reactions of trimethylborate (TMB) in a FTICR can be used to differentiate the ortho-, meta-, and para-isomers of protonated aromatic compounds, such as those formed during degradation of lignin. All three regioisomers could be differentiated for methoxyphenols and hydroxyphenols. However, only the differentiation of the ortho-isomer from the meta- and para-isomers was possible for hydroxyacetophenones and hydroxybenzoic acids. Consideration of the previously reported proton affinities at all basic sites in the isomeric hydroxyphenols, and the calculated proton affinities at all basic sites in the three methoxyphenol isomers, revealed that the proton affinities of the analytes relative to that of TMB play an important role in determining whether and how they react with TMB. The loss of two methanol molecules (instead of one) from the adducts formed with TMB either during ion-molecule reactions, or during sustained-off resonance irradiated collision-activated dissociation of the ion-molecule reaction products, revealed the presence of two functionalities in almost all the isomers. This finding supports earlier results suggesting that TMB can be used to count the functionalities in unknown oxygen-containing analytes.
Sigma-Aldrich
Trimethyl borate, purum, ≥99.0% (GC)
Sigma-Aldrich
Trimethyl borate, purified by redistillation, ≥99.5%
Sigma-Aldrich
Trimethyl borate, ≥98%
Sigma-Aldrich
Trimethyl borate, 99.999% (trace metal basis)