MilliporeSigma
  • Sultone opening with [18F]fluoride: an efficient 18F-labelling strategy for PET imaging.
Sultone opening with [18F]fluoride: an efficient 18F-labelling strategy for PET imaging.
Chemical communications (Cambridge, England) (2011-09-29)
Sébastien Schmitt, Cédric Bouteiller, Louisa Barré, Cécile Perrio
ABSTRACT
Sultones were subject to ring opening by nucleophilic attack with [(18)F]fluoride to afford easily purified (18)F-labelled hydrophilic sulfonated products in high yields. A two-step sequence including radiofluorination and coupling to lysine was then developed from a bis-sultone precursor as a model approach for the labelling of biopolymers.