MilliporeSigma
  • Carboxylesterase expression in human dental pulp cells: role in regulation of BisGMA-induced prostanoid production and cytotoxicity.
Carboxylesterase expression in human dental pulp cells: role in regulation of BisGMA-induced prostanoid production and cytotoxicity.
Acta biomaterialia (2011-09-29)
M-C Chang, L-D Lin, F-H Chuang, C-P Chan, T-M Wang, J-J Lee, P-Y Jeng, W-Y Tseng, H-J Lin, J-H Jeng
ABSTRACT
Biocompatibility of dentin bonding agents (DBA) and composite resin may affect the treatment outcome (e.g., healthy pulp, pulpal inflammation, pulp necrosis) after operative restoration. Bisphenol-glycidyl methacrylate (BisGMA) is one of the major monomers present in DBA and resin. Prior studies focused on salivary esterase for metabolism and degradation of resin monomers clinically. This study found that human dental pulp cells expressed mainly carboxylesterase-2 (CES2) and smaller amounts of CES1A1 and CES3 isoforms. Exposure to BisGMA stimulated CES isoforms expression of pulp cells, and this event was inhibited by catalase. Exogenous addition of porcine esterase prevented BisGMA- and DBA-induced cytotoxicity. Interestingly, inhibition of CES by bis(p-nitrophenyl) phosphate (BNPP) and CES2 by loperamide enhanced the cytotoxicity of BisGMA and DBA. Addition of porcine esterase or N-acetyl-l-cysteine prevented BisGMA-induced prostaglandin E(2) (PGE(2)) and PGF(2α) production. In contrast, addition of BNPP and loperamide, but not mevastatin, enhanced BisGMA-induced PGE(2) and PGF(2α) production in dental pulp cells. These results suggest that BisGMA may induce the cytotoxicity and prostanoid production of pulp cells, leading to pulpal inflammation or necrosis via reactive oxygen species production. Expression of CES, especially CES2, in dental pulp cells can be an adaptive response to protect dental pulp against BisGMA-induced cytotoxicity and prostanoid release. Resin monomers are the main toxic components in DBA, and the ester group is crucial for monomer toxicity.
Sigma-Aldrich
Bis(4-nitrophenyl) phosphate, 99%
Sigma-Aldrich
Bis(p-nitrophenyl) phosphate sodium salt, ≥99% (TLC)