MilliporeSigma
Substrate stereoselectivity of mammalian D-aspartyl endopeptidase.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-09-29)
Tadatoshi Kinouchi, Norihiko Fujii, Noriko Fujii
ABSTRACT
The formation and accumulation of D-aspartate residue (D-Asp) in proteins caused by oxidative stress leads to dysfunction and/or denaturation of proteins, and is consequently responsible for aging-related misfolding diseases such as cataracts, prion disease, and Alzheimer's disease. We sought to identify that an unknown protease selectively degrades the noxious D-Asp-containing protein, namely D-aspartyl endopeptidase (DAEP), and finally purified it from the inner mitochondrial membrane of mouse liver. In order to analyze the substrate stereoselectivity of DAEP, we synthesized a peptide corresponding to 55-65 (Thr-Val-Leu-Asp-Ser-Gly-Ile-Ser-Glu-Val-Arg) of human αA-crystallin and its corresponding diastereoisomers in which L-α-Asp was replaced with L-β-, D-α- or D-β-Asp residue at position 58. Following incubation of that peptide with purified DAEP, it was only degraded at D-α-Asp(58), independent of ATP or NAD. This result indicates that DAEP stereoselectively recognizes and degrades its substrate at the internal D-α-Asp residue. DAEP therefore seems to physiologically serve as the quality control system against the noxious D-Asp-containing protein in the long life span of mammals.
Sigma-Aldrich
D-Aspartic acid, ReagentPlus®, 99%