MilliporeSigma
  • Antioxidant and lipid peroxidation activities in rats fed with Aspergillus carbonarius carotenoid.
Antioxidant and lipid peroxidation activities in rats fed with Aspergillus carbonarius carotenoid.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-09-20)
Anbarasu Kumar, Akshatha Hosahalli Srikanta, S P Muthukumar, Umesh-Kumar Sukumaran, Vijayalakshmi Govindaswamy
ABSTRACT
Effect of feeding partially saturated canthaxanthin (PSC), purified from Aspergillus carbonarius mutant, was studied using four groups of female albino rats (n=6) for 4 weeks. While the control group received basal diet ad libitum, Groups I, II and III were fed with basal diet containing 50, 100 and 250 ppm PSC, respectively. PSC feeding did not cause any significant changes in food intake and there was no gain in body weight either. PSC included in the diet significantly decreased cholesterol in blood. There was 44.75% and 60.54% decrease in LDL-cholesterol in rats fed with 50 and 100 ppm carotenoid. Hepatic ascorbic acid content increased by 44.59% in rats fed with 50 ppm PSC. Dietary PSC at 250 ppm lowered lipid peroxides by 19.49%. Activities of antioxidant enzymes, glutathione transferase and catalase were significantly higher in serum and liver of PSC fed rats compared to the controls. The results suggested that PSC feeding can induce hypocholesterolmic and antioxidant properties in rats.
Product Description
Supelco
Canthaxanthin (trans), analytical standard