MilliporeSigma
  • Bioactive metabolites from biotransformation of paeonol by the white-rot basidiomycete Coriolus versicolor.
Bioactive metabolites from biotransformation of paeonol by the white-rot basidiomycete Coriolus versicolor.
Natural product communications (2011-09-20)
Xiao-Jun Li, Xin-Wei Shi, Qi Shuai, Jin-Ming Gao, An-Ling Zhang
ABSTRACT
Biotransformation of paeonol (1) with the white-rot basidiomycete Coriolus versicolor afforded two metabolites, 2,4-dihydroxyacetophenone (2) and 2,5-dihydroxy-4-methoxyacetophenone (3), which were identified by spectroscopic methods. Compound 3 showed higher antioxidative, antibacterial, antifungal activities than 1 or 2. The results demonstrate for the first time that C. versicolor has the capacities to catalyze hydroxylation and demethylation reactions on the aromatic compound.
Sigma-Aldrich
2′-Hydroxy-4′-methoxyacetophenone, 99%
Sigma-Aldrich
2′,4′-Dihydroxyacetophenone, 99%