MilliporeSigma
  • [Photocatalytic degradation of 1,2, 4-trichlorobenzene with TiO2 coated on carbon nanotubes].
[Photocatalytic degradation of 1,2, 4-trichlorobenzene with TiO2 coated on carbon nanotubes].
Huan jing ke xue= Huanjing kexue (2011-09-20)
Wei Zhang, Zhou Shi, Qian Zhang, Hu Zhang, Shun-Kai Xu
ABSTRACT
Effects and kinetics of photocatalytic degradation of 1,2,4-trichlorobenzene with composite photocatalysts of TiO2 coated on carbon nanotubes and nano-TiO2 photocatalysts were studied, respectively. It was found that the composite photocatalysts was better than the nano-TiO2 for photocatalytic degradation of 1,2,4-trichlorobenzene. After 60 min UV irradiation at 254 nm, there was 70% of the initial 1,2,4-trichlorobenzene degraded with the composite photocatalyst while only 51% of the initial 1,2,4-trichlorobenzene degraded with the nano-TiO2. Results of photocatalytic degradation kinetics also proved that the 1,2,4-trichlorobenzene degradation rate constant with the composite photocatalyst was 0.019 6 h(-1), which was increased by 63.3% compared to that with nano-TiO2. According to molecular structure theory, analysis of dissociation energy of C--Cl bond of chlorobenzenes predicated that main pathway of photocatalytic degradation of 1,2,4-trichlorobenzene followed that its ortho-Cl could be took off first to form 1, 4-dichlorobenzene, and it might be further dechlorized and finally be mineralized. The first step of photodechlorination for 1,2,4-trichlorobenzene was also experimentally confirmed through identified 1,4-dichlorobenzene as its product.
Sigma-Aldrich
1,2,4-Trichlorobenzene, anhydrous, ≥99%
Supelco
1,2,4-Trichlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
1,2,4-Trichlorobenzene, suitable for HPLC, ≥99%
Sigma-Aldrich
1,2,4-Trichlorobenzene, ReagentPlus®, ≥99%