MilliporeSigma
A chromatin immunoprecipitation protocol for small cell numbers.
Methods in molecular biology (Clifton, N.J.) (2011-09-14)
Philippe Collas
ABSTRACT
Chromatin immunoprecipitation (ChIP) is a widely used technique to get a snap-shot of protein-DNA interactions in cells. ChIP has notably been used for mapping the location of modified histones, transcription factors, or chromatin remodeling enzymes in the genome, often in relation to transcription or differentiation. Conventional ChIP protocols however, have for a long time required large numbers of cells, which has limited the applicability of ChIP to rare or small cell samples. In recent years, ChIP assays for small cell numbers (in the 10,000-100,000 range) have been recently reported by us and others. This chapter describes a micro (μ)ChIP procedure for multiple parallel ChIPs from a single chromatin batch from 1,000 cells.
Sigma-Aldrich
3-Methyl-1-butanol, ACS reagent, ≥98.5%
Sigma-Aldrich
3-Methyl-1-butanol, reagent grade, 98%
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Supelco
3-Methylbutanol, analytical standard
Sigma-Aldrich
3-Methylbutanol, for molecular biology, BioReagent, ≥98.5%
Sigma-Aldrich
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit