MilliporeSigma
  • Malabaricone C from Myristica cinnamomea exhibits anti-quorum sensing activity.
Malabaricone C from Myristica cinnamomea exhibits anti-quorum sensing activity.
Journal of natural products (2011-09-14)
Yee Meng Chong, Wai Fong Yin, Chia Yong Ho, Mohamad Rais Mustafa, A Hamid A Hadi, Khalijah Awang, Putri Narrima, Chong-Lek Koh, David R Appleton, Kok-Gan Chan
ABSTRACT
A methanol-soluble extract of the bark of Myristica cinnamomea was found to exhibit anti-quorum sensing activity, and subsequent bioassay-guided isolation led to the identification of the active compound malabaricone C (1). Compound 1 inhibited violacein production by Chromobacterium violaceum CV026 when grown in the presence of a cognate signaling molecule, N-3-oxohexanoyl-homoserine lactone. Furthermore, 1 inhibited the quorum sensing-regulated pyocyanin production and biofilm formation in Pseudomonas aeruginosa PAO1. These results suggest that the anti-quorum sensing activity of 1 and related molecules should be investigated further.
Sigma-Aldrich
Violacein from Janthinobacterium lividum, >98% (violacein (minimum 85% violacein) and deoxyviolacein, HPLC)