MilliporeSigma
  • Sensitive detection of esculetin based on a CdSe nanoparticles-decorated poly(diallyldimethylammonium chloride)-functionalized graphene nanocomposite film.
Sensitive detection of esculetin based on a CdSe nanoparticles-decorated poly(diallyldimethylammonium chloride)-functionalized graphene nanocomposite film.
The Analyst (2011-09-08)
Daban Lu, Yan Zhang, Shaoxiong Lin, Letao Wang, Chunming Wang
ABSTRACT
An electrochemical sensor based on a CdSe nanoparticles (NPs)-decorated poly(diallyldimethylammonium chloride) (PDDA)-functionalized graphene (CdSe-PDDA-G) nanocomposite was fabricated for the sensitive detection of esculetin. The nanocomposite was characterized by X-ray diffraction (XRD), ultraviolet/visible spectra (UV-vis) and transmission electron microscopy (TEM). Cyclic voltammetry (CV) and differential pulse voltammetry (DPV) were used to investigate the electrochemical behaviors of esculetin on the CdSe-PDDA-G composite film-modified glassy carbon electrode (GCE). The experimental results indicated that the incorporation of CdSe NPs with PDDA-G greatly enhanced the electrochemical response of esculetin. This electrochemical sensor displayed satisfactory analytical performance for esculetin detection over a range from 1.0 × 10(-8) to 5.0 × 10(-5) mol L(-1) with a detection limit of 4.0 × 10(-9) mol L(-1) (S/N = 3). Moreover, the sensor also exhibited good reproducibility and stability, and could be used for the detection of esculetin in real samples with satisfactory results.
Sigma-Aldrich
6,7-Dihydroxycoumarin, 98%