MilliporeSigma
  • Larvicidal activity of (-)-dihydroguaiaretic acid derivatives against Culex pipiens.
Larvicidal activity of (-)-dihydroguaiaretic acid derivatives against Culex pipiens.
Bioscience, biotechnology, and biochemistry (2011-09-08)
Hisashi Nishiwaki, Ayaka Hasebe, Yuya Kawaguchi, Miki Akamatsu, Yoshihiro Shuto, Satoshi Yamauchi
ABSTRACT
The larvicidal activity against Culex pipiens of all stereoisomers of dihydroguaiaretic acid (DGA) and secoisolariciresinol was measured, and these DGAs were found to be potent. Sixteen (-)-DGA derivatives were then newly synthesized to analyze their structure-activity relationship. Two derivatives monohydroxylated at the 3- or 4-position of the 7-phenyl group of DGA induced acute paralytic activity in the mosquitoes. Derivatives with several hydroxyl groups had lower activity than the natural compound, suggesting that hydrophobicity was probably an important factor for their insecticidal activity.
Sigma-Aldrich
Secoisolariciresinol, ≥95.0% (HPLC)