MilliporeSigma
Nitric oxide-releasing ruthenium nanoparticles.
Chemical communications (Cambridge, England) (2011-09-03)
Chi-Ming Ho, Kai-Jun Liao, Chun-Nam Lok, Chi-Ming Che
ABSTRACT
Nitric oxide-releasing ruthenium nanoparticles were synthesized by the reaction of alkanethiolate-protected ruthenium nanoparticles with tert-butyl nitrite ((t)BuONO), and their water-soluble derivatives are able to deliver NO to proteins such as reduced myoglobin upon light irradiation in aqueous media.
Sigma-Aldrich
Butyl nitrite, 95%