MilliporeSigma
  • Comparison of performance of the novel chromogenic spectra VRE agar to that of bile esculin azide and Campylobacter agars for detection of vancomycin-resistant enterococci in fecal samples.
Comparison of performance of the novel chromogenic spectra VRE agar to that of bile esculin azide and Campylobacter agars for detection of vancomycin-resistant enterococci in fecal samples.
Journal of clinical microbiology (2011-09-02)
S G Jenkins, L Raskoshina, A N Schuetz
ABSTRACT
A total of 142 stool specimens were evaluated for vancomycin-resistant enterococcus (VRE). Twenty-four-hour sensitivities and specificities, respectively, were 98% and 95% for Spectra VRE chromogenic agar (Remel, Lenexa, KS), 86% and 92% for bile esculin azide with vancomycin (BEAV; Remel), and 96.5% and 92% for Campylobacter agar (CAMPY; Remel). Spectra VRE and CAMPY are significantly more sensitive at 24 h than BEAV.
Sigma-Aldrich
Esculin hydrate, ≥98.0% (HPLC)
Sigma-Aldrich
Esculin hydrate, ≥98%