MilliporeSigma
  • Combining SERCA2a activation and Na-K ATPase inhibition: a promising new approach to managing acute heart failure syndromes with low cardiac output.
Combining SERCA2a activation and Na-K ATPase inhibition: a promising new approach to managing acute heart failure syndromes with low cardiac output.
Discovery medicine (2011-09-01)
Mihai Gheorghiade, Andrew P Ambrosy, Mara Ferrandi, Patrizia Ferrari
ABSTRACT
Heart failure (HF) patients are a medically complex and heterogeneous population with multiple cardiac and non-cardiac comorbidities. Although there are a multitude of etiologic substrates and initiating and amplifying mechanisms contributing to disease progression, these pathophysiologic processes ultimately all lead to impaired myocardial function. The myocardium must both pump oxygenated, nutrient-rich blood throughout the body (systolic function) and receive deoxygenated, nutrient-poor blood returning from the periphery (diastolic function). At the molecular level, it is well-established that Ca2+ plays a central role in excitation-contracting coupling with action potentials stimulating the opening of L-type Ca2+ in the plasma membrane and ryanodine receptor 2 (RyR2) in the sarcoplasmic reticulum (SR) membrane during systole and the Na-Ca2+ exchanger and SERCA2a returning Ca2+ to the extracellular space and SR, respectively, during diastole. However, there is increasing recognition that impaired Ca2+ cycling may contribute to myocardial dysfunction. Preclinical studies and clinical trials indicate that combining SERCA2a activation and Na-K ATPase inhibition may increase contractility (inotropy) and facilitate active relaxation (lusitropy), improving both systolic and diastolic functions. Istaroxime, a novel luso-inotrope that activates SERCA2a and inhibits the Na-K ATPase, is currently in phase II clinical development and has been shown to improve systolic and diastolic functions and central hemodynamics, increase systolic but not diastolic blood pressure, and decrease substantially heart rate. Irrespective of its clinical utility, the development of istaroxime has evolved our understanding of the clinical importance of inhibiting the Na-K ATPase in order to obtain a clinically significant effect from SERCA2a activation in the setting of myocardial failure.