MilliporeSigma
Photoinduced oxidation of the insecticide phenothrin on soil surfaces.
Journal of agricultural and food chemistry (2011-09-01)
Yusuke Suzuki, Andrea Lopez, Marian Ponte, Takuo Fujisawa, Luis O Ruzo, Toshiyuki Katagi
ABSTRACT
Photodegradation profiles of the pyrethroid insecticide phenothrin on a moistened U.S. soil thin layer was investigated by using its predominant component, the 1R-trans-isomer (I), under continuous exposure to light at >290 nm from a xenon arc lamp. Its degradation was moderately accelerated by irradiation with half-lives of 5.7-5.9 days (dark control 21-24 days), mainly via successive oxidation of the 2-methylprop-1-enyl group and ester cleavage followed by mineralization to carbon dioxide. Spectroscopic and cochromatographic analyses showed that the major degradates were the alcohol and ketone derivatives of I formed via photoinduced oxidation of the 2-methylprop-1-enyl group by singlet oxygen. The photoinduced generation of singlet oxygen in/on the soil surface was confirmed by using chemical trapping reactions together with ESR spectroscopy.
Product Description
Supelco
Phenothrin, PESTANAL®, analytical standard