MilliporeSigma
  • Thyrotropin (TSH) regulates triiodothyronine (T3) production in the unicellular Tetrahymena.
Thyrotropin (TSH) regulates triiodothyronine (T3) production in the unicellular Tetrahymena.
Acta biologica Hungarica (2011-08-16)
G Csaba, Eva Pállinger
ABSTRACT
The aim of the experiments was to study the regulation of triiodothyronine (T3) production in the unicellular Tetrahymena. Untreated and troph-hormone treated specimen were prepared and in different timepoints T3 content was measured and compared by immunocytochemical flow cytometry. 0.1 or 0.001 IU TSH in tryptone-yeast medium stimulated T3 synthesis at 10, 20, 30 min, but does not stimulate after 1 h. The overlapping gonadotropic hormone (GTH) also did it, however only at 10 min. In Losina salt solution (physiological for Tetrahymena) the effect was weaker, however outer amino acid source was not absolutely needed for the production of the hormone. The results show that the TSH regulation of thyroid hormone synthesis (storage, secretion) and troph-hormone overlap can be deduced to a unicellular level. This may allow the hypothesis that the endocrine mechanisms proved at a low level of phylogeny are preserved for the higher ranked organisms.
Product Description
Millipore
Proteose Peptone, Enzymatic hydrolysate
Millipore
Peptone from milk solids, Refined hydrolysate
Millipore
Peptone from casein and other animal proteins, suitable for microbiology
Millipore
Peptone from animal tissue, from meat
Sigma-Aldrich
Peptone from animal tissue, from meat, BioReagent, suitable for cell culture, suitable for plant cell culture
Millipore
Peptone Primatone® RL, suitable for microbiology