MilliporeSigma
  • Understanding of the mass spectrometric fragmentation pathways of a few potentially genotoxic haloaniline isomers in their protonated form by collision-induced dissociation.
Understanding of the mass spectrometric fragmentation pathways of a few potentially genotoxic haloaniline isomers in their protonated form by collision-induced dissociation.
Journal of pharmaceutical and biomedical analysis (2011-08-16)
Rex Patrick, N S Nagarajan, Hefeng Pan, Subhra Kanti Saha, Jayan Rapai, Venkatarao Torlikonda, Manoranjan Panda, Sulur G Manjunatha, Sudhir Nambiar
ABSTRACT
Collision-induced dissociation (CID) mass spectra of a few haloaniline isomers, (chloroanilines, dichloroanilines, difluoroanilines, chloro-fluoroanilines and bromo-fluoroanilines) were characterized. The mass spectral behaviour of difluoroanilines was different from those of the corresponding regioisomers of the other haloanilines. For all ortho regioisomers except difluoroanilines, CID mass spectra resulted in hydrogen halide as well as halogen radical loss. In the case of difluoroanilines, peaks corresponding to hydrogen fluoride loss were observed during the same process. Meta and para-haloanilines have the tendency to lose either ammonia or halogen radicals. Six regioisomers of dichloroanilines were subjected to hydrogen/deuterium exchange experiments in solution to determine the CID fragmentation pathways. From the experimental results we propose two fragmentation pathways for the dicholoroanilines: (a) formation of aza-biheterocyclic intermediate and (b) via heterolytic hydrogen transfer from the charged center. The demonstrated unique characteristics in CID mass fragmentation of haloanilines may be useful in identification and differentiation of isomers as impurities during chemical process development. A good use of the ortho effect is the significant differentiation between 2-chloro-4-fluoroaniline and 4-chloro-2-fluoroaniline by CID mass spectra.
Sigma-Aldrich
4-Fluoroaniline, 99%
Supelco
2-Fluoroaniline, ≥99%