MilliporeSigma
  • Multiple resonance heteronuclear decoupling under MAS: dramatic increase of spectral resolution at moderate magnetic field and MAS frequencies.
Multiple resonance heteronuclear decoupling under MAS: dramatic increase of spectral resolution at moderate magnetic field and MAS frequencies.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2011-08-16)
Charlotte Martineau, Franck Engelke, Francis Taulelle
ABSTRACT
The effects of multiple-resonance heteronuclear decoupling under magic angle spinning (MAS) on the resolution of one-dimensional (19)F and (31)P and various two-dimensional MAS NMR spectra and on the residual non-refocusable coherence lifetimes in fluorinated aluminophosphate AlPO(4)-CJ2, i.e. a compound that contains numerous highly abundant nuclei but no homonuclear spin bath, has been investigated. The design of the four-channel ((1)H, (19)F, (27)Al, (31)P) MAS probe used for this study is first described. (1)H and (1)H-(27)Al double-resonance decouplings allows lengthening the optimized transverse relaxation T(2)(opt) and increasing the resolution in the (19)F and (31)P dimensions. Under the application of multi-nuclear decoupling, a two-dimensional (19)F-(31)P CP-HETCOR correlation spectrum for AlPO(4)-CJ2 is recorded with unprecedented high-resolution in the two dimensions. Moreover, because (1)H-decoupling increases the (19)F T(2)(opt), it has been applied during the entire duration of the 2D NMR experiments, allowing the direct use of residual small interactions to generate (19)F-(19)F and (19)F-(27)Al 2D NMR correlation spectra in AlPO(4)-CJ2.
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Aluminum phosphate, reagent grade
Sigma-Aldrich
Aluminum phosphate monobasic, purum, ≥95.0% (gravimetric)
Sigma-Aldrich
Aluminum phosphate monobasic solution, ~50% in H2O