MilliporeSigma
  • Synthesis and vibrational spectroscopic analysis of some Hofmann type clathrates.
Synthesis and vibrational spectroscopic analysis of some Hofmann type clathrates.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-08-09)
Cemal Parlak, Mustafa Şenyel
ABSTRACT
New Hofmann type clathrates in the form of M(pp)(2)Ni(CN)(4)·2G (where pp = 1-phenylpiperazine, G = 1,4-dioxane and M = Ni, Co or Cd) have been prepared in powder form and their FT-IR and FT-Raman spectra are reported. The results suggest that the present compounds are similar in structure to the Hofmann type clathrates and their structures consist of conjugated polymeric layers of |M-Ni(CN)(4)|(∞) with the pp bound to the metal (M) atom.
Sigma-Aldrich
1-Phenylpiperazine, ≥97.0%