MilliporeSigma
  • Photodegradation of (-)-epigallocatechin-3-gallate in topical cream formulations and its photostabilization.
Photodegradation of (-)-epigallocatechin-3-gallate in topical cream formulations and its photostabilization.
Journal of pharmaceutical and biomedical analysis (2011-08-03)
Anna Bianchi, Nicola Marchetti, Santo Scalia
ABSTRACT
The aim of the study was to examine the photostability of the major catechin of green tea, (-)-epigallocatechin-3-gallate (EGCG), which possesses important antioxidant and skin photoprotective properties. In order to simulate realistic conditions of use of topical preparations, the photolysis studies were performed in model creams (oil-in-water emulsions) containing 1% (w/w) EGCG and exposed to a solar simulator at an irradiance corresponding to natural sunlight. The extent of photodegradation was measured by HPLC-UV and HPLC-ESI-MS. EGCG was found to decompose by 68.9±2.3%, after 1h irradiation. Addition of the coantioxidants, vitamin E or butylated hydroxytoluene to the emulsion formulation, significantly enhanced the photolability of the catechin, the EGCG loss reached 85.7±1.3% and 80.5±1.4%, respectively. On the other hand, inclusion of the UVB (290-320nm) filter, ethylhexyl methoxycinnamate in the cream produced a small but significant reduction of EGCG photodegradation to 61.0±2.9%, while the UVA (320-400nm) filter, butyl methoxydibenzoylmethane was ineffective (EGCG degradation, 67.8±1.5%). A more marked decrease in the light-induced decomposition of EGCG to 51.6±2.7% was achieved, under the same conditions, using the water-soluble UVB filter, benzophenone-4 (BP-4). This effect was concentration dependent, maximal EGCG photostabilization (catechin loss, 29.4±2.2%) was attained in the presence of 2.1% (w/w) BP-4. Therefore, BP-4 represents a useful additive to improve the light stability of EGCG in topical formulations for skin photoprotection.
Product Description
Supelco
Sulisobenzone, analytical standard
Sigma-Aldrich
5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid, ≥97.0% (HPLC)