MilliporeSigma
  • Synthesis of N-[2-(2,4-Difluorophenoxy)trifluoromethyl-3-pyridyl]sulfonamides and their inhibitory activities against secretory phospholipase A₂.
Synthesis of N-[2-(2,4-Difluorophenoxy)trifluoromethyl-3-pyridyl]sulfonamides and their inhibitory activities against secretory phospholipase A₂.
Chemical & pharmaceutical bulletin (2011-08-02)
Hitoshi Nakayama, Keiichi Ishihara, Satoshi Akiba, Jun'ichi Uenishi
ABSTRACT
N-[2-(2,4-Difluorophenoxy)trifluoromethyl-3-pyridyl]sulfonamide derivatives 3-6 were prepared by the reaction of 3-pyridylamines and sulfonyl chlorides. Inhibitory activities of these compounds toward secretory phospholipase A₂ (sPLA₂) were examined and N-[2-(2,4-difluorophenoxy)-5-trifluoromethyl-3-pyridyl]-2-naphthalenesulfonamide (5c) was found to be the most potent against sPLA₂ with an IC₅₀ value of 90 µM.
Sigma-Aldrich
Trifluoromethanesulfonamide, 95%