MilliporeSigma
  • Study of serum interaction with a cationic nanoparticle: Implications for in vitro endocytosis, cytotoxicity and genotoxicity.
Study of serum interaction with a cationic nanoparticle: Implications for in vitro endocytosis, cytotoxicity and genotoxicity.
International journal of pharmaceutics (2011-08-02)
Maysaloun Merhi, Christophe Youta Dombu, Alizée Brient, Jiang Chang, Anne Platel, Frank Le Curieux, Daniel Marzin, Fabrice Nesslany, Didier Betbeder
ABSTRACT
We used well-characterized and positively charged nanoparticles (NP(+)) to investigate the importance of cell culture conditions, specifically the presence of serum and proteins, on NP(+) physicochemical characteristics, and the consequences for their endocytosis and genotoxicity in bronchial epithelial cells (16HBE14o-). NP(+) surface charge was significantly reduced, proportionally to NP(+)/serum and NP(+)/BSA ratios, while NP(+) size was not modified. Microscopy studies showed high endocytosis of NP(+) in 16HBE14o-, and serum/proteins impaired this internalization in a dose-dependent manner. Toxicity studies showed no cytotoxicity, even for very high doses of NP(+). No genotoxicity was observed with classic comet assay while primary oxidative DNA damage was observed when using the lesion-specific repair enzyme, formamidopyrimidine DNA-glycosylase (FPG). The micronucleus test showed NP(+) genotoxicity only for very high doses that cannot be attained in vivo. The low toxicity of these NP(+) might be explained by their high exocytosis from 16HBE14o- cells. Our results confirm the importance of serum and proteins on nanoparticles endocytosis and genotoxicity.
Sigma-Aldrich
Glycidyltrimethylammonium chloride, technical, ≥90% (calc. based on dry substance, AT)