MilliporeSigma
  • The anti-metastatic efficacy of β-ionone and the possible mechanisms of action in human hepatocarcinoma SK-Hep-1 cells.
The anti-metastatic efficacy of β-ionone and the possible mechanisms of action in human hepatocarcinoma SK-Hep-1 cells.
The British journal of nutrition (2011-07-27)
Chin-Shiu Huang, Shih-Chieh Lyu, Jen-Yin Chen, Miao-Lin Hu
ABSTRACT
β-Ionone (BI), a precursor for carotenoids, is widely distributed in fruit and vegetables. Recent in vitro studies have demonstrated the potential anti-metastatic effects of BI, but the mechanisms underlying such actions are not clear. Because liver cancer is the most endemic cancer in Taiwan and in a large region of the world, we hereby investigate the anti-metastatic effects of BI and its mechanisms of actions in a highly metastatic human hepatocarcinoma SK-Hep-1 cells. We show that incubation of cells with BI (1-50 μm) for 24 and 48 h significantly inhibited cell invasion, migration and adhesion. Mechanistically, incubation of cells with BI (1-50 μm) for 24 h resulted in the following: (1) significant inhibition of matrix metalloproteinase (MMP)-2, MMP-9 and urokinase-type plasminogen activator activities, (2) up-regulation of protein expression of the tissue inhibitor of matrix metalloproteinase (TIMP)-1, TIMP-2 and plasminogen activator inhibitor-1, (3) down-regulation of the expression of migration-related proteins, including focal adhesion kinase (FAK), phosphorylated form of FAK, Rho, Rac1 and Cdc42 and (4) up-regulation of the expression of nm23-H1 protein (P < 0·05). Overall, the results show that BI effectively inhibits the metastasis of SK-Hep-1 cells, and this effect involves the regulation of gene expression and signal pathways related to invasion and migration.
Sigma-Aldrich
β-Ionone, natural, ≥95%, FG
Sigma-Aldrich
β-Ionone, predominantly trans, ≥97%, FCC, FG
Sigma-Aldrich
β-Ionone, 96%