MilliporeSigma
  • Wet chemical synthesis and self-assembly of SnS2 nanoparticles on TiO2 for quantum dot-sensitized solar cells.
Wet chemical synthesis and self-assembly of SnS2 nanoparticles on TiO2 for quantum dot-sensitized solar cells.
Journal of nanoscience and nanotechnology (2011-07-23)
Hiroki Tsukigase, Yoshikazu Suzuki, Marie-Hélène Berger, Takashi Sagawa, Susumu Yoshikawa
ABSTRACT
SnS2 nanoparticles were synthesized through a simple wet chemical process at room temperature. The SnS2 nanoparticles were approximately spherical in shape and had diameter about 3-4 nm. SnS2-sensitized TiO2 electrodes were fabricated by the immersion of chemically modified TiO2 to well-dispersed SnS2 solution for 72 h (i.e., self-assembly method.) SnS2-sensitized TiO2 electrodes were applied in quantum dot-sensitized solar cells (QDSSCs). Under AM1.5 irradiation with 100 mW/cm2 light intensity (at 1 sun), the short-circuit current density (J(sc)), the open-circuit voltage (V(oc)), the fill factor (FF), and the energy conversion efficiency (eta) were 0.47 mA/cm2, 0.29 V, 0.58 and 0.081%, respectively.