MilliporeSigma
  • Engineered polymer-media interfaces for the long-term self-renewal of human embryonic stem cells.
Engineered polymer-media interfaces for the long-term self-renewal of human embryonic stem cells.
Biomaterials (2011-07-22)
Elizabeth F Irwin, Rohini Gupta, Derek C Dashti, Kevin E Healy
ABSTRACT
We have developed a synthetic polymer interface for the long-term self-renewal of human embryonic stem cells (hESCs) in defined media. We successfully cultured hESCs on hydrogel interfaces of aminopropylmethacrylamide (APMAAm) for over 20 passages in chemically-defined mTeSR™1 media and demonstrated pluripotency of multiple hESC lines with immunostaining and quantitative RT-PCR studies. Results for hESC proliferation and pluripotency markers were both qualitatively and quantitatively similar to cells cultured on Matrigel™-coated substrates. Mechanistically, it was resolved that bovine serum albumin (BSA) in the mTeSR™1 media was critical for cell adhesion on APMAAm hydrogel interfaces. This study uniquely identified a robust long-term culture surface for the self-renewal of hESCs without the use of biologic coatings (e.g., peptides, proteins, or Matrigel™) in completely chemically-defined media that employed practical culturing techniques amenable to clinical-scale cell expansion.
Sigma-Aldrich
Methacrylamide, 98%