MilliporeSigma
  • Phenylpropanoid and phenylisoprenoid metabolites from Asteraceae species as inhibitors of protein carbonylation.
Phenylpropanoid and phenylisoprenoid metabolites from Asteraceae species as inhibitors of protein carbonylation.
Phytochemistry (2011-07-19)
Marta Marín, Rosa M Giner, M Carmen Recio, Salvador Máñez
ABSTRACT
Three phenolic antioxidant and anti-inflammatory compounds: 7-methylaromadendrin, isoprenylhydroquinone glucoside, and 3.5-dicaffeoylquinic acid methyl ester, all isolated from Western Mediterranean Asteraceae species, have been studied for their inhibitory activity against protein carbonylation, a harmful post-translational modification of peptide chains associated with degenerative diseases. All compounds have proven to be effective, with 50% inhibitory concentration (IC₅₀) values in the micromolar range, against bovine serum albumin carbonylation caused by hypochlorite, peroxynitrite, and phorbol ester-induced leukocyte oxidative burst.