MilliporeSigma
  • Surface-plasmon-coupled emission of Rhodamine 110 in a silica nanolayer.
Surface-plasmon-coupled emission of Rhodamine 110 in a silica nanolayer.
Chemphyschem : a European journal of chemical physics and physical chemistry (2011-07-14)
Simeonika Rangełowa-Jankowska, Dawid Jankowski, Beata Grobelna, Ignacy Gryczyński, Zygmunt Gryczyński, Robert Bogdanowicz, Piotr Bojarski
ABSTRACT
The first observation of strong directional surface-plasmon-coupled emission (SPCE) of Rhodamine 110 in silica nanofilms deposited on silver nanolayers is reported. The preparation of the material is described in detail. The intensity of SPCE exceeds 10 times that of free space fluorescence and total linear light polarization in the SPCE ring is observed. A new experimental setup and an original data collection method is presented. Our material completely preserves its fluorescence properties for at least eight months.
Sigma-Aldrich
Rhodamine 110 chloride, Dye content ≥75 %
Sigma-Aldrich
Rhodamine 110 chloride, suitable for fluorescence, BioReagent, ≥99.0% (UV)