MilliporeSigma
  • Free energy calculations offer insights into the influence of receptor flexibility on ligand-receptor binding affinities.
Free energy calculations offer insights into the influence of receptor flexibility on ligand-receptor binding affinities.
Journal of computer-aided molecular design (2011-07-08)
Jožica Dolenc, Sereina Riniker, Roberto Gaspari, Xavier Daura, Wilfred F van Gunsteren
ABSTRACT
Docking algorithms for computer-aided drug discovery and design often ignore or restrain the flexibility of the receptor, which may lead to a loss of accuracy of the relative free enthalpies of binding. In order to evaluate the contribution of receptor flexibility to relative binding free enthalpies, two host-guest systems have been examined: inclusion complexes of α-cyclodextrin (αCD) with 1-chlorobenzene (ClBn), 1-bromobenzene (BrBn) and toluene (MeBn), and complexes of DNA with the minor-groove binding ligands netropsin (Net) and distamycin (Dist). Molecular dynamics simulations and free energy calculations reveal that restraining of the flexibility of the receptor can have a significant influence on the estimated relative ligand-receptor binding affinities as well as on the predicted structures of the biomolecular complexes. The influence is particularly pronounced in the case of flexible receptors such as DNA, where a 50% contribution of DNA flexibility towards the relative ligand-DNA binding affinities is observed. The differences in the free enthalpy of binding do not arise only from the changes in ligand-DNA interactions but also from changes in ligand-solvent interactions as well as from the loss of DNA configurational entropy upon restraining.
Sigma-Aldrich
Netropsin dihydrochloride, from Streptomyces netropsis, ≥98% (HPLC and TLC), powder
Sigma-Aldrich
Bromobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Bromobenzene, ≥99.5% (GC)