MilliporeSigma
  • Assessment of a bone biopsy technique for measuring tiludronate in horses: a preliminary study.
Assessment of a bone biopsy technique for measuring tiludronate in horses: a preliminary study.
Canadian journal of veterinary research = Revue canadienne de recherche veterinaire (2011-07-07)
Catherine Delguste, Michèle Doucet, Annick Gabriel, Jérôme Guyonnet, Olivier M Lepage, Hélène Amory
ABSTRACT
This study assessed the feasibility of measuring tiludronate in horses using a minimally invasive bone biopsy technique. Eight horses were treated with intravenous (IV) tiludronate [1 mg/kg bodyweight (BW)], either once (n = 4) or twice, 28 d apart (n = 4). The horses that were treated once were euthanized on days 1, 43, 57, or 92 and those that were treated twice, were euthanized on days 112, 154, 194, or 364. Bone samples were taken bilaterally from each horse at 4 sites: the third metacarpal bone (MCIII), the 13th rib (R13), the tuber coxae (TC), and the cuboid bone (CB). Test samples were taken with a 5-mm diameter dental drill, while larger reference samples were taken with an osteotome. The concentrations of tiludronate were measured by high performance liquid chromatography (HPLC) with ultraviolet (UV) detection. The TC was the easiest site to sample, and no technical difficulties were encountered for extraction and measurement. Drill sampling at the MCIII was difficult. Moreover, both the extraction and measurement caused technical problems and results were unreliable in most cases (93%). Drill samples obtained from the R13 were very small and access to the CB required considerable dissection, which would not be feasible in vivo. Forty-six percent and 36% of the tiludronate measurements performed on the R13 and CB samples, respectively, were unreliable. The ratio of tiludronate concentrations ranged from 73% to 185% (median: 118%) in the TC, 65% to 208% (median: 81%) in the R13, and 26% to 110% (median: 57%) in the CB. In all but 1 horse, the highest concentrations of tiludronate were found in the TC. It was concluded that bone biopsies performed at the TC were adequate for measuring tiludronate in horses and should be considered in future for repeated measurements over time in living animals.
Sigma-Aldrich
Tiludronate disodium salt hydrate, ≥98% (HPLC)