MilliporeSigma
  • Antifungal activity of 2 lactic acid bacteria of the Weissella genus isolated from food.
Antifungal activity of 2 lactic acid bacteria of the Weissella genus isolated from food.
Journal of food science (2011-07-07)
Dora Ndagano, Thibaut Lamoureux, Carine Dortu, Sophie Vandermoten, Philippe Thonart
ABSTRACT
In the present study, a total of 116 lactic acid bacteria (LAB) strains isolated from Mill flour and fermented cassava were screened for their antifungal activity. Three strains among 116 were selected for their strongest inhibitory activity against food molds. These 3 strains were Lactobacillus plantarum VE56, Weissella cibaria FMF4B16, and W. paramesenteroides LC11. The compounds responsible for the antifungal activity were investigated. The strains displayed an inhibitory activity against targeted molds at acidic pH. However, the influence of organic acids was rejected according to the calculated minimal inhibitory concentration (MIC). Antifungal compounds were investigated in the cell-free supernatants and phenyllactic acid (PLA) was detected in different amounts with a maximal concentration for Lb. plantarum VE56 (0.56 mM). Hydroxy fatty acid, such as 2-hydroxy-4-methylpentanoic acid, was also produced and involved in the inhibitory activity of Lb. plantarum VE56 and W. paramesenteroides LC11. Antifungal LAB are known to produce PLA and 3-hydroxy fatty acids and other organic acids with antifungal activity. This short communication focuses on antifungal activity from Weissella genus. The antifungal activity was attributed to antifungal compounds identified such as PLA, 2-hydroxy-4-methylpentanoic acid, and other organic acids. Nevertheless, the concentration produced in the cell-free supernatant was too low to compare to their MIC, suggesting that the inhibitory activity was caused by a synergy of these different compounds. Antifungal LAB are interesting to prevent food spoilage in fermented food and prolong their shelf life. In this way, chemical preservatives could be avoided and replaced by natural preservatives.
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%
Sigma-Aldrich
D-(+)-3-Phenyllactic acid, 98%
Sigma-Aldrich
3-Phenyllactic acid, ≥98%