MilliporeSigma
  • Adhesion promotion via noncovalent interactions in self-healing polymers.
Adhesion promotion via noncovalent interactions in self-healing polymers.
ACS applied materials & interfaces (2011-07-07)
Gerald O Wilson, Mary M Caruso, Stuart R Schelkopf, Nancy R Sottos, Scott R White, Jeffrey S Moore
ABSTRACT
Dimethylnorbornene ester (DNE) is successfully used as a noncovalent adhesion promoter. DNE was confirmed to copolymerize with dicyclopentadiene (DCPD) to yield a copolymer with better adhesion to an EPON 828 epoxy matrix relative to poly(DCPD) alone. The mechanical properties of the copolymer were comparable to that of poly(DCPD) alone. An optimized blend of the monomers was encapsulated using a urea-formaldheyde microencapsulation procedure and the resulting capsules were used for in situ self-healing experiments. Improved healing efficiency was observed for samples containing the DCPD/DNE capsules under conditions in which the monomers were efficiently polymerized.
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer