MilliporeSigma
  • A simple LC-MS/MS method to determine plasma and cerebrospinal fluid levels of albendazole metabolites (albendazole sulfoxide and albendazole sulfone) in patients with neurocysticercosis.
A simple LC-MS/MS method to determine plasma and cerebrospinal fluid levels of albendazole metabolites (albendazole sulfoxide and albendazole sulfone) in patients with neurocysticercosis.
Biomedical chromatography : BMC (2011-07-02)
Iliana González-Hernández, María Isabel Ruiz-Olmedo, Graciela Cárdenas, Helgi Jung-Cook
ABSTRACT
The development and validation of an LC-MS/MS method for the simultaneous determination of albendazole metabolites (albendazole sulfoxide and albendazole sulfone) in human plasma are described. Samples of 200 μL were extracted with ether-dichloromethane-chloroform (60:30:10, v/v/v). The chromatographic separation was performed using a C(18) column with methanol-formic acid 20 mmol/L (70:30) as the mobile phase. The method was linear in a range of 20-5000 ng/mL for albendazole sulfoxide and 10-1500 ng/mL for albendazole sulfone. For both analytes the method was precise (RSD < 12%) and accurate (RE <7%) with high recovery (>90%). The method was successfully applied to determine the plasma and cerebrospinal fluid levels of albendazole sulfoxide and albendazole sulfone in patients with subarachnoidal neurocysticercosis who received albendazole at 30 mg/kg per day for 7 days. This LC-MS/MS method yielded a quick, simple and reliable protocol for determining albendazole sulfoxide and albendazole sulfone concentrations in plasma and cerebrospinal fluid samples and is applicable to therapeutic monitoring.
Product Description
Supelco
Albendazole sulfoxide, VETRANAL®, analytical standard