MilliporeSigma
  • Concentrations and environmental risk of chlorothalonil in air near potato fields in Prince Edward Island, Canada.
Concentrations and environmental risk of chlorothalonil in air near potato fields in Prince Edward Island, Canada.
Pest management science (2011-06-29)
Christine Garron, Bill Ernst, Gary Julien, René Losier, Kimberly Davis
ABSTRACT
Chlorothalonil is used extensively in Prince Edward Island for the control of blight on potatoes, and has been measured throughout summer months in ambient air monitoring studies. This study was designed to characterize near-field drift of chlorothalonil, and to evaluate how exposure to measured air concentrations might impact upon local wildlife species. Air samples were collected using high-volume samplers at three distances downwind of sprayed potato fields, and at four different time intervals. Chlorothalonil was detected in 73% of prespray samples (from < 0.013 to 0.440 µg m(-3)). The mean air concentration 0 m away from the field edge during spray was 5.8 µg m(-3), which was 7-41 times higher than concentrations measured in similar studies. Air concentrations were found to decline exponentially both with distance and time, although 0 m and 30 m concentrations at 2 h post-spray remained significantly (3 times) higher than prespray (P < 0.05). Wind speed was not found to be significantly correlated with air concentration; however, increases in temperature were found to be significantly correlated with increases in air concentrations at all sample times, at 0 m and 30 m distances. Frequent prespray detections and high post-spray concentrations suggest that both chronic and acute exposures to non-target species are occurring; however, risk calculations suggest a relatively low risk of effects to representative species. One measurement showed that a human-health-based short-term guideline had been exceeded, indicating that potential impacts to humans from chlorothalonil under current application conditions cannot be discounted.
Product Description
Supelco
Chlorothalonil, PESTANAL®, analytical standard