MilliporeSigma
  • Effect of ethanolic extract of Rubia peregrina L. (Rubiaceae) on monoamine-mediated behaviour.
Effect of ethanolic extract of Rubia peregrina L. (Rubiaceae) on monoamine-mediated behaviour.
Natural product research (2011-06-29)
Sanjay Kasture, Anupama Pawar, Ameya Kasture, Caterina Foddis, Maria Assunta Frau, Andrea Maxia
ABSTRACT
In the traditional system of medicine in Sardinia, Rubia peregrina (RP) is reported as an aphrodisiac herb. Since aphrodisiacs may also have dopaminergic activity, and there can be a reciprocal relationship between dopaminergic and serotonergic functions in the central nervous system, the aim here was to study the effect of the ethanolic extract of the aerial parts of R. peregrina on monoamine-mediated behaviour. The RP (100 and 200 mg kg(-1) intraperitoneally) significantly inhibited haloperidol (1 mg kg(-1) i.p.) induced catalepsy in mice (p < 0.01, dopamine-mediated response) lithium sulphate induced head twitches in rats (serotonin-mediated response) and was without effect on clonidine-induced hypothermia in rats (noradrenaline-mediated response). The study concludes that R. peregrina improves the dopaminergic function, diminishes the serotonin-mediated function and is devoid of any effect on the noradrenergic function. The study suggests that we should carry out further experiments to investigate the psychopharmacological profile of R. peregrina.
Sigma-Aldrich
Lithium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium sulfate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Lithium sulfate, ≥98.5% (titration)