MilliporeSigma
  • Enzymatic probing of model lipid membranes: phospholipase A2 activity toward monolayers modified by oxicam NSAIDs.
Enzymatic probing of model lipid membranes: phospholipase A2 activity toward monolayers modified by oxicam NSAIDs.
The journal of physical chemistry. B (2011-06-21)
Katarzyna Czapla, Beata Korchowiec, Monika Orlof, Jenifer Rubio Magnieto, Ewa Rogalska
ABSTRACT
Three nonsteroidal anti-inflammatory oxicam drugs, namely meloxicam, piroxicam, and tenoxicam, were used to modify the properties of monomolecular films formed with 1,2-dilauroyl-sn-glycero-3-phosphocholine, 1,2-dilauroyl-sn-glycero-3-phosphoethanolamine, or 1,2-dilauroyl-sn-glycero-3-phospho-(1-rac-glycerol). These systems were examined via surface pressure and surface electrical potential measurements, polarization modulation infrared reflection absorption spectroscopy, and Brewster angle microscopy. Moreover, phospholipase A2 activity was used to differentiate between the three drugs. Our results reveal that the oxicams studied modify membrane properties, namely hydration of the lipid polar heads, orientation of the molecules, and morphology of the domains. Phospholipase A2 was shown to be sensitive to the presence of the drugs in the systems studied; the activity of the enzyme correlates with the effect of meloxicam, piroxicam, and tenoxicam on the monolayer properties. The latter indicates that the anti-inflammatory action of oxicams may be related to interference with phospholipase activity in addition to cyclooxygenase inhibition.
Product Description
Tenoxicam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tenoxicam, NSAID
Avanti
12:0 PG, Avanti Research - A Croda Brand