MilliporeSigma
  • Spectroscopic studies on interaction and sonodynamic damage of metallochlorophyllin (Chl-M (M=Fe, Zn and Cu)) to protein under ultrasonic irradiation.
Spectroscopic studies on interaction and sonodynamic damage of metallochlorophyllin (Chl-M (M=Fe, Zn and Cu)) to protein under ultrasonic irradiation.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-06-18)
Jingqun Gao, Zhiqiu Wang, Jun Wang, Xudong Jin, Yuwei Guo, Kai Li, Ying Li, Pingli Kang
ABSTRACT
In this paper, the chlorophyll derivatives, metallochlorophyllin (Chl-M) (M=Fe, Zn and Cu) including chlorophyllin iron (Chl-Fe), chlorophyllin zinc (Chl-Zn) and chlorophyllin copper (Chl-Cu), were adopted as sonosensitizers to combine with ultrasonic irradiation, and the sonodynamic damage of bovine serum albumin (BSA) was investigated. At first, the interaction of Chl-M with BSA was studied by fluorescence spectroscopy. The results show that the quenching mechanism belongs to a static process and among them the affinity of Chl-Fe to BSA is the most obvious. Then, some influence factors on the sonodynamic damage of BSA molecules in the presence of Chl-M under ultrasonic irradiation were also studied. Synchronous fluorescence spectra show that the binding and damage sites of Chl-M to BSA molecule are mainly on the tryptophan (Trp) residues. The generation of ROS in Chl-M sonodynamic process is estimated by the method of Oxidation-Extraction Spectrometry (OEP). This paper may offer some valuable references for the study of the sonodynamic activity of Chl-M and the effect of the central metals. Synchronously, it contributes to the application of Chl-M in SDT for tumor treatment.
Sigma-Aldrich
Chlorophyllin sodium copper salt