MilliporeSigma
  • Amphiphilic DNA block copolymers: nucleic acid-polymer hybrid materials for diagnostics and biomedicine.
Amphiphilic DNA block copolymers: nucleic acid-polymer hybrid materials for diagnostics and biomedicine.
Methods in molecular biology (Clifton, N.J.) (2011-06-16)
Jan Zimmermann, Minseok Kwak, Andrew J Musser, Andreas Herrmann
ABSTRACT
DNA-polymer conjugates have been recognized as versatile functional materials in many different fields ranging from nanotechnology to diagnostics and biomedicine. They combine the favorable properties of nucleic acids and synthetic polymers. Moreover, joining both structures with covalent bonds to form bioorganic hybrids allows for the tuning of specific properties or even the possibility of evolving completely new functions. One important class of this type of material is amphiphilic DNA block copolymers, which, due to microphase separation, can spontaneously adopt nanosized micelle morphologies with a hydrophobic core and a DNA corona. These DNA nano-objects have been explored as vehicles for targeted gene and drug delivery, and also as programmable nanoreactors for organic reactions. Key to the successful realization of these potential applications is that (1) DNA block copolymer conjugates can be fabricated in a fully automated fashion by employing a DNA synthesizer; (2) hydrophobic compounds can be loaded within their interior; and (3) they can be site-specifically functionalized by a convenient nucleic acid hybridization procedure. This chapter aims to broaden the range of biodiagnostic and biomedical applications of these materials by providing a comprehensive outline of the preparation and characterization of multifunctional DNA-polymer nanoparticles.
Sigma-Aldrich
Poly(propylene glycol), average Mn ~1,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,700
Sigma-Aldrich
Poly(propylene glycol), average Mn ~725
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~4,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~425
Sigma-Aldrich
Polypropylene glycol, P 2,000
Sigma-Aldrich
Polypropylene glycol, P 400